missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Red Flange Plug Caps, 17 mm

Catalog No. 02681530
Change view
Click to view available options
Color:
Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
02681530 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02681530 Supplier Fisherbrand™ Supplier No. 6263RPLFH

These polyethylene (PE) flanged plug caps provide exceptional leak-resistance in test tubes, centrifuge tubes and round cuvettes.

  • Two flexible flanges that press firmly against the inside wall of the tubes, thereby providing better leak-resistance than standard plug caps.

Specifications

Material Polyethylene
Color Red
Quantity 1000/Cs.
For Use With (Equipment) 17 mm Tubes and 15 mL Centrifuge Tubes