missing translation for 'onlineSavingsMsg'
Learn More

Red-CLA [Chemiluminescence Reagent], TCI America™

Catalog No. a53111mg
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A53111MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A53111MG Supplier TCI America Supplier No. A53111MG

Chemical Identifiers

CAS 886840-56-8
Molecular Formula C50H51N7O9S2
Molecular Weight (g/mol) 958.118
InChI Key USCHMYZSZWGIDF-UHFFFAOYSA-N
Synonym [2-[4-[4-[3,7-Dihydro-2-methyl-3-oxoimidazo[1,2-a]pyrazin-6-yl]phenoxy]butyramido]ethylamino]sulforhodamine 101
PubChem CID 90477284
SMILES CC1=NC2=CNC(=CN2C1=O)C3=CC=C(C=C3)OCCCC(=O)NCCNS(=O)(=O)C4=CC(=C(C=C4)S(=O)(=O)[O-])C5=C6C=C7CCC[N+]8=C7C(=C6OC9=C1CCCN2C1=C(CCC2)C=C59)CCC8

Specifications

Color Black-Purple
Quantity 1 mg
Formula Weight 958.12
Physical Form Crystalline Powder
Chemical Name or Material Red-CLA [Chemiluminescence Reagent]
TSCA No
Recommended Storage Freezer