accessibility menu, dialog, popup

UserName

CAS RN 886840-56-8

IUPAC Name: 16-[5-({2-[4-(4-{2-methyl-3-oxo-3H,7H-imidazo[1,2-a]pyrazin-6-yl}phenoxy)butanamido]ethyl}sulfamoyl)-2-sulfonatophenyl]-3-oxa-9λ⁵,23-diazaheptacyclo[17.7.1.1⁵,⁹.0²,¹⁷.0⁴,¹⁵.0²³,²⁷.0¹³,²⁸]octacosa-1,4,9(28),13,15,17,19(27)-heptaen-9-ylium
Synonyms: 16-[5-({2-[4-(4-{2-methyl-3-oxo-7H-imidazo[1,2-a]pyrazin-6-yl}phenoxy)butanamido]ethyl}sulfamoyl)-2-sulfonatophenyl]-3-oxa-9λ⁵,23-diazaheptacyclo[17.7.1.1⁵,⁹.0²,¹⁷.0⁴,¹⁵.0²³,²⁷.0¹³,²⁸]octacosa-1,4,9(28),13,15,17,19(27)-heptaen-9-ylium
Molecular Weight (g/mol): 958.12
Molecular Formula: C50H51N7O9S2
InChi Key: USCHMYZSZWGIDF-UHFFFAOYSA-N
SMILES: CC1=NC2=CNC(=CN2C1=O)C1=CC=C(OCCCC(=O)NCCNS(=O)(=O)C2=CC(=C(C=C2)S([O-])(=O)=O)C2=C3C=C4CCC[N+]5=C4C(CCC5)=C3OC3=C4CCCN5CCCC(C=C23)=C45)C=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results