missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO Recycled Aluminum Sample Pans

Catalog No. 01670165
Change view
Click to view available options
Diameter (Metric):
96 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
01670165 96 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01670165 Supplier METTLER TOLEDO Supplier No. 31058332

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Single-use aluminum sample pans with a minimum of 88% recycled content for moisture and dry content determination.

  • Single-use, oil- and fat-free surface
  • Each carton contains 80 sample pans
  • Compatible with all Mettler Toledo moisture analyzers

Specifications

Autoclavable Autoclavable
Bottom Shape Flat
Color Silver
Disposable Single-use
Material Aluminum
Shape Round
Sterility Non-sterile
Type Moisture Pan
For Use With (Equipment) Infrared Moisture Analyzers
Quantity 80 Pk.
Diameter (English) 3.75 in.
Diameter (Metric) 96 mm
Show More Show Less