Learn More
Fisherbrand™ Rectangular Cover Glasses
Made of the finest optical borosilicate glass, with uniform thickness and size.
Supplier: Fisherbrand™ 24X301602811T
Description
These cover glasses feature a high-spectral transmission quality and finely adaptive refractive index suited for microscopy. Sold by weight, comes in a pack of 1 oz.
- Made of colorless borosilicate glass
- Available in a variety of thicknesses and sizes
Chemical Identifiers
| Rectangle | |
| Borosilicate Glass | |
| Microscopy, Histology, Cytology, Hematology, Microbiology | |
| 1 oz./Pk. |
Specifications
| Coverslip | |
| Borosilicate Glass | |
| 30 x 24 mm | |
| 1 oz./Pk. |
| Rectangle | |
| 0.13 to 0.17 mm (#1) | |
| Microscopy, Histology, Cytology, Hematology, Microbiology |