Learn More
Fisherbrand™ Rectangular Cover Glasses
Made of the finest optical borosilicate glass, with uniform thickness and size.
Supplier:  Fisherbrand™ 22X501.5602811T
 View more versions of this product
Catalog No. 12541024CA
Only null left
Description
Description
These cover glasses feature a high-spectral transmission quality and finely adaptive refractive index suited for microscopy. Sold by weight, comes in a pack of 1 oz.
  • Made of colorless borosilicate glass
  • Available in a variety of thicknesses and sizes
Specifications
Chemical Identifiers
Rectangle
Borosilicate Glass
Microscopy, Histology, Cytology, Hematology, Microbiology
1 oz./Pk.
Specifications
Coverslip
Borosilicate Glass
50 x 22 mm
1 oz./Pk.
Rectangle
0.16 to 0.19 mm (#1.5)
Microscopy, Histology, Cytology, Hematology, Microbiology
Product Suggestions
Product Suggestions