missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RECQ4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103642
Description
Antibody detects endogenous levels of total RECQL4.
The protein encoded by this gene is a DNA helicase that belongs to the RecQ helicase family. DNA helicases unwind double-stranded DNA into single-stranded DNAs and may modulate chromosome segregation. This gene is predominantly expressed in thymus and testis. Mutations in this gene are associated with Rothmund-Thomson, RAPADILINO and Baller-Gerold syndromes.Specifications
| RECQ4 | |
| Polyclonal | |
| Unconjugated | |
| RECQL4 | |
| ATP-dependent DNA helicase Q4; DNA helicase, RecQ-like type 4; DNA helicase, RecQ-like, type 4; RecQ helicase protein-like 4; RecQ helicase-like 4; RecQ like helicase 4; RecQ protein-like 4; Recq4; RECQL4; RTS | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 9401 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O94761 | |
| RECQL4 | |
| A synthesized peptide derived from human RECQL4(Accession O94761), corresponding to amino acid residues G231-Q281. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |