missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novex™ Recombinant Protein G
Recombinant Protein G
$256.00 - $624.00
Specifications
| Conjugate | Unconjugated |
|---|---|
| Form | Powder |
| Protein Subtype | Protein G |
| Content And Storage | 2-8°C |
| Shipping Condition | Wet Ice |
Description
Recombinant protein G is produced in a strain of E. coli which contains the gene for protein G from Streptococcus sp. It is supplied as a salt-free lyophilized powder.Recombinant protein G binds to the Fc region of many types of immunoglobulin G including:
Recombinant protein G does not bind to IgG from chicken or cat, or to human IgA, IgM, or serum albumin. It binds weakly to IgG from dog.
Specifications
| Unconjugated | |
| Protein G | |
| Wet Ice |
| Powder | |
| 2-8°C |
For Research Use Only. Not for use in diagnostic procedures.