missing translation for 'onlineSavingsMsg'
Learn More

Novex™ Recombinant Protein A-Sepharose™ 4B

Catalog No. p-7076200
Encompass
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
101141 5 mL
101142 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 101141 Supplier Novex™ Supplier No. 101141
Only null left

Recombinant Protein A-Sepharose™ 4B

This product is prepared by covalently coupling highly purified Staph. aureus Cowan strain Protein A produced in E. Coli to Sepharose™ 4B.

This product is supplied as a 50% suspension consisting of recombinant Protein A-Sepharose™ 4B and PBS (10 mM phosphate-buffered saline, pH 7.4, containing 0.1% ).

Applications: Immunoglobulin Affinity Purification, Immunoprecipitation

Specifications

Product Type Protein A Sepharose
Column Type Affinity
Stationary Phase Protein A
Content And Storage 2°C to 8°C. Do Not Freeze.
This product is supplied as a 50% suspension (5 ml Recombinant Protein A-Sepharose™ 4B + 5 ml PBS) Wet Ice
Quantity 5 mL
Form Suspension

For Research Use Only. Not for use in diagnostic procedures.