missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human HO-2/HMOX2 Protein

Catalog No. NBP137089
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP137089 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP137089 Supplier R&D Systems™ Supplier No. NBP137089

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems. Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-264 of Human HO-2/HMOX2 The Recombinant Human HO-2/HMOX2 Protein is derived from E. coli. The Recombinant Human HO-2/HMOX2 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_002125
Concentration 1 mg/mL
For Use With (Application) SDS-PAGE
Formulation 20 mM Tris-HCl buffer (pH 8.0), 10% glycerol, 1 mM DTT
Gene ID (Entrez) 3163
Molecular Weight (g/mol) M.W. (theoretical): 30.5 kDa
Name HO-2/HMOX2 Protein
Purification Method Protein
Quantity 0.1 mg
Immunogen MSAEVETSEG VDESEKKNSG ALEKENQMRM ADLSELLKEG TKEAHDRAEN TQFVKDFLKG NIKKELFKLA TTALYFTYSA LEEEMERNKD HPAFAPLYFP MELHRKEALT KDMEYFFGEN WEEQVQCPKA AQKYVERIHY IGQNEPELLV AHAYTRYMGD LSGGQVLKKV AQRALKLPST GEGTQFYLFE NVDNAQQFKQ LYRARMNALD LNMKTKERIV EEANKAFEYN MQIFNELDQA GSTLARETLE DGFPVHDGKG DMRK
Storage Requirements Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.)
Gene Symbol HMOX2
Cross Reactivity Human
Product Line Novus
Purity or Quality Grade >90%, by SDS-PAGE
Protein HO-2/HMOX2
Show More Show Less