missing translation for 'onlineSavingsMsg'
Learn More

Manganese Sulfate, Monohydrate, Reagent, ACS, 98-101%, Spectrum™ Chemical

Catalog No. 18604220
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Poly Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604220 50 kg Poly Bottle
18604214 125 g Poly Bottle
18604215 500 g Poly Bottle
18604217 2.5 kg Poly Bottle
18604216 6 x 500 g Poly Bottle
18604218 4 x 2.5 kg Poly Bottle
18604219 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18604220 Supplier Spectrum Chemical Mfg Cor Supplier No. M111550KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

M1115, 10034-96-5, MnSO4·H2O

Spectrum™ Chemical Manganese Sulfate, Monohydrate, Reagent, ACS , is an inorganic compound that like many metal sulfates can form a variety of hydrates such as heptahydrates, monohydrates, tetrahydrates, pentahydrates. When these salts are dissolved, they tend to give a light pink solution, a characteristic of manganese sulfate. As an ACS grade Reagent, Spectrum Chemical manufactured manganese sulfate is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 10034-96-5
Molecular Formula H2MnO5S
Molecular Weight (g/mol) 169.01
MDL Number MFCD00149159
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
IUPAC Name manganese(2+) hydrate sulfate
SMILES O.[Mn++].[O-]S([O-])(=O)=O

Specifications

Physical Form Fine Crystals or Crystalline Powder
CAS Max % 100%
Quantity 50 kg
Loss on Ignition 10 to 13%
Packaging Poly Bottle
Percent Purity 98 to 101%
Grade ACS/Reagent
Chemical Name or Material Manganese Sulfate, Monohydrate