missing translation for 'onlineSavingsMsg'
Learn More

Rebaudioside A 98.0+%, TCI America™

Catalog No. p-7135693
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
R00955G 5 g
R009525G 25 g
2 options

Catalog No. R00955G

Supplier: TCI America R00955G

Only null left

Chemical Identifiers

CAS 58543-16-1
Molecular Formula C44H70O23
Molecular Weight (g/mol) 967.02
MDL Number MFCD02183463
InChI Key HELXLJCILKEWJH-UHFFFAOYNA-N
PubChem CID 131676233
IUPAC Name 3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl 13-{[5-hydroxy-6-(hydroxymethyl)-3,4-bis({[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy})oxan-2-yl]oxy}-5,9-dimethyl-14-methylidenetetracyclo[11.2.1.0¹,¹⁰.0⁴,⁹]hexadecane-5-carboxylate
SMILES CC12CCCC(C)(C1CCC13CC(=C)C(C1)(CCC23)OC1OC(CO)C(O)C(OC2OC(CO)C(O)C(O)C2O)C1OC1OC(CO)C(O)C(O)C1O)C(=O)OC1OC(CO)C(O)C(O)C1O

Specifications

Melting Point 235°C
Color White
Quantity 5 g
Formula Weight 967.02
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Rebaudioside A
RTECSNumber NZ8174800
TSCA No