missing translation for 'onlineSavingsMsg'
Learn More

Rebamipide 98.0+%, TCI America™

Catalog No. p-7135687
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Catalog No. Quantity
R00851G 1 g
R00855G 5 g
R008525G 25 g
3 options

Catalog No. R00851G

Supplier: TCI America R00851G

Only null left

Chemical Identifiers

CAS 90098-04-7
Molecular Formula C19H15ClN2O4
Molecular Weight (g/mol) 370.79
MDL Number MFCD00866895,MFCD11114396
InChI Key ALLWOAVDORUJLA-UHFFFAOYNA-N
Synonym 2-(4-Chlorobenzamido)-3-[2(1H)-quinolinon-4-yl]propionic Acid
PubChem CID 5042
IUPAC Name 2-[(4-chlorophenyl)formamido]-3-(2-oxo-1,2-dihydroquinolin-4-yl)propanoic acid
SMILES OC(=O)C(CC1=CC(=O)NC2=CC=CC=C12)NC(=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point 290°C
Color White
Quantity 1 g
Formula Weight 370.79
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Rebamipide
RTECSNumber VC2518500
TSCA No