missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Edvotek™ Reagents for Electrophoresis: UltraSpec-Agarose™
Edvotek™ Reagents for Electrophoresis: UltraSpec-Agarose™ DNA Electrophoresis Grade reagent.
$814.33 - $814.33
Specifications
| Color | White |
|---|---|
| Format | Bottle |
| For Use With (Equipment) | Mini Gel Tank |
| Product Type | Agarose |
| Separation of | DNA |
Description
DNA Electrophoresis GradeSpecifications
| White | |
| Mini Gel Tank | |
| DNA | |
| Agarose or Polyacrylamide Gel electrophoresis |
| Bottle | |
| Agarose | |
| Electrophoresis | |
| Agarose |