Learn More
Thermo Scientific™ Reagent Reservoirs
GREENER_CHOICE
Catalog No. 04100517
Change view
Click to view available options
Capacity (Metric):
25 mL
Material:
Polystyrene
Sterility:
Non-sterile
Packaging:
10 Bags of 10
Bulk
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Material Sterility Packaging
04100517 25 mL Polystyrene Non-sterile 10 Bags of 10
04100518 25 mL Polystyrene Non-sterile Bulk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 04100517 Supplier Thermo Scientific™ Supplier No. 95128095
Choose these reagent reservoirs, designed to provide a variety of solutions to suit all of your pipetting procedures.
Thermo Scientific™ Reagent Reservoirs provide a variety of solutions to suit all of your pipetting procedures.
Innovation goes beyond our pipetters and tips. Even our reagent reservoirs have been designed to provide solutions tailored to your procedures, rather than the other way around. Four variations of reservoirs are offered, 25mL, 25mL with divider, 100mL and a disposable polystyrene 75mL reservoir.
Features:
  • Trough within a trough maximizes the amount of liquid accessible to pipette tips when using small amounts of reagent
  • Pour-off spouts on all four corners reduce spillage when pouring reagents out of reservoir
  • Graduations on inside wall enable quick measurement of remaining liquid
  • Extra-wide base adds rigidity and stability to reservoir, helping to avoid spills
  • 25mL divided reservoir allows the pipetting of two different reagents with up to eight channels on one side, and up to four on the other; maximizes recovery if using only single channel and small amount of reagent
Specifications
Color White
Capacity (Metric) 25 mL
Material Polystyrene
Sterility Non-sterile
Type Reagent Reservoir with Divider
Packaging 10 Bags of 10
Disposable Single-use