missing translation for 'onlineSavingsMsg'
Learn More

Simport™ Scientific Reagent Reservoirs

For labs involved with fraction collecting, compound storage, plate replication, and mixing applications. Suitable for a range of liquid handling configurations from 8 or 12-channel pipettes to 96 or 384-pipette heads. Ideal for small-volume robotic tips used in high-throughput instruments.

Supplier:  Simport™ Scientific R2255

 View more versions of this product

Catalog No. 01670663


Only null left
Explore available promotions

Description

Description

  • Robot friendly and designed for simple, economical handling of reagents in automated systems
  • Conforms to ANSI/SLAS standard dimensions
  • Compatible with virtually any automated liquid handler, stacker, and manual procedure
  • Bottom designed to enable funneling of liquid, maximizing reagent recovery with pipettes
  • Low-profile, chemical and heat resistant
  • Can be stored at temperatures as low as -80°C
  • Can be gamma irradiated
  • Certified DNase-, RNase- and pyrogen-free
  • Made from natural virgin polypropylene
Specifications

Specifications

Yes
15 mL
Polypropylene
10 Pk.
No
Flat
Clear
127.8 x 85.5 x 31.4 mm
Non-sterile
50 Cs.
Single-use
12 Wells
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions