missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Reagent Reservoir, Bulk wrap
Description
- Superior durability with enhanced walls and crush-resistant turrets at the corners
- Easy handling with tabs to prevent stacked reservoirs from sticking together
Specifications
Specifications
| Capacity (Metric) | 50 mL |
| Dimensions (L x W x H) | 4.5 x 1.1 x 1.4 in. |
| Material | PVC |
| Sterility | Non-sterile |
| Type | Reagent Reservoir |
| Packaging | Bulk |
| Quantity | 100 |
| Autoclavable | Not Autoclavable |
| Channels | 1 |
| Disposable | Single-use |
| Show More |