missing translation for 'onlineSavingsMsg'
Learn More
Learn More
- Home
- Products
- Tanks
- Reservoirs and Liners
- Fisherbrand™ Reagent Reservoir
Fisherbrand™ Reagent Reservoir
Catalog No. 01670461

Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
55 mL
100 mL
Sterility:
Non-sterile
Sterile
Packaging:
5 Pk. Wrap
Bulk
Individually Wrapped
Material:
Polystyrene
PVC
17 product options available for selection
| Catalog No. | Capacity (Metric) | Sterility | Packaging | Material |
|---|---|---|---|---|
| 01670461 | 10 mL | Sterile | Individually Wrapped | Polystyrene |
| 14127052 | 25 mL | Sterile | 5 Pk. Wrap | Polystyrene |
| 14127057 | 50 mL | Sterile | 5 Pk. Wrap | Polystyrene |
| 14127054 | 50 mL | Sterile | 5 Pk. Wrap | PVC |
| 14127064 | 100 mL | Sterile | 5 Pk. Wrap | Polystyrene |
| 14127058 | 50 mL | Sterile | Individually Wrapped | Polystyrene |
| 14127062 | 25 mL | Sterile | Individually Wrapped | Polystyrene |
| 14127065 | 100 mL | Sterile | Individually Wrapped | Polystyrene |
| 14127072 | 55 mL | Sterile | Individually Wrapped | Polystyrene |
| 14127067 | 100 mL | Non-sterile | Bulk | PVC |
| 14127074 | 50 mL | Non-sterile | Bulk | PVC |
| 14127060 | 55 mL | Non-sterile | Bulk | Polystyrene |
| 14127066 | 100 mL | Non-sterile | Bulk | Polystyrene |
| 14127063 | 25 mL | Non-sterile | Bulk | Polystyrene |
| 14127053 | 50 mL | Non-sterile | Bulk | Polystyrene |
| 14127061 | 55 mL | Non-sterile | Bulk | PVC |
| 01670454 | 10 mL | Non-sterile | Bulk | Polystyrene |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
Description
- Superior durability with enhanced walls and crush-resistant turrets at the corners
- Easy handling with tabs to prevent stacked reservoirs from sticking together
Specifications
Specifications
| Capacity (English) | 0.34 oz. |
| Certifications/Compliance | DNase, RNase, and Pyrogen free quality certificate available upon request, Traceability Certificate available upon request |
| Color | White |
| Capacity (Metric) | 10 mL |
| Material | Polystyrene |
| Sterility | Sterile |
| Type | Reagent Reservoir |
| For Use With (Application) | Pipetting procedures |
| Packaging | Individually Wrapped |
| Quantity | 100 Pk. |
| Autoclavable | Not Autoclavable |
| Channels | 1 |
| Disposable | Single-use |
| Bottom Shape | V-bottom |
| Show More Show Less |