Learn More
Fisherbrand™ Reagent Reservoir
Catalog No. 14127063
Change view
Click to view available options
Capacity (Metric):
25 mL (10 and 15 mL divided)
10 mL
25 mL
50 mL
55 mL
100 mL
Sterility:
Non-sterile
Sterile
Packaging:
5 Pk. Wrap
Bulk
Individually Wrapped
Material:
Polypropylene
Polystyrene
PVC
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Sterility Packaging Material
14127063 25 mL Non-sterile Bulk Polystyrene
14127055 25 mL (10 and 15 mL divided) Sterile 5 Pk. Wrap Polypropylene
14127052 25 mL Sterile 5 Pk. Wrap Polystyrene
14127057 50 mL Sterile 5 Pk. Wrap Polystyrene
14127054 50 mL Sterile 5 Pk. Wrap PVC
14127064 100 mL Sterile 5 Pk. Wrap Polystyrene
14127058 50 mL Sterile Individually Wrapped Polystyrene
14127056 25 mL (10 and 15 mL divided) Sterile Individually Wrapped Polystyrene
14127062 25 mL Sterile Individually Wrapped Polystyrene
14127065 100 mL Sterile Individually Wrapped Polystyrene
14127072 55 mL Sterile Individually Wrapped Polystyrene
14127067 100 mL Non-sterile Bulk PVC
14127074 50 mL Non-sterile Bulk PVC
14127060 55 mL Non-sterile Bulk Polystyrene
14127066 100 mL Non-sterile Bulk Polystyrene
14127053 50 mL Non-sterile Bulk Polystyrene
14127061 55 mL Non-sterile Bulk PVC
01670461 10 mL Sterile Individually Wrapped Polystyrene
01670454 10 mL Non-sterile Bulk Polystyrene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
Catalog No. 14127063 Supplier Fisherbrand™ Supplier No. 120642F2
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures.
  • Superior durability with enhanced walls and crush-resistant turrets at the corners
  • Easy handling with tabs to prevent stacked reservoirs from sticking together
Specifications
Capacity (Metric) 25 mL
Dimensions (L x W x H) 4.4 x 0.9 x 1.1 in.
Material Polystyrene
Sterility Non-sterile
Type Reagent Reservoir
Packaging Bulk
Quantity 100
Autoclavable Not Autoclavable
Channels 1
Disposable Single-use
Bottom Shape V-bottom
Show More Show Less