missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Reagent Reservoir
GREENER_CHOICE

Catalog No. 01670454
Change view
Click to view available options
Capacity (Metric):
10 mL
Sterility:
Non-sterile
Sterile
Packaging:
Bulk Wrapped
Individually Wrapped
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Sterility Packaging
01670454 10 mL Non-sterile Bulk Wrapped
01670461 10 mL Sterile Individually Wrapped
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01670454 Supplier Fisherbrand™ Supplier No. 120638F2
Only null left

Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures.

  • Superior durability with enhanced walls and crush-resistant 'turrets' at the corners
  • Easy handling with tabs to prevent stacked reservoirs from sticking together
  • Graduation markings on the top edge and inner walls allows measurements of liquid to be easily identified and reduces wastage of expensive reagents
  • Pour-off Spouts conveniently located on all four corners enables easy spill-free pouring and reduces chance of dripping
  • Divided reservoir design allows pipetting of two different reagents or maximizes recovery if using only single channel and small amount of reagent
  • Manufactured and gamma irradiated in an ISO certified facility
  • Certified DNase, RNase and pyrogen free
  • Packaged individually in an easy open HDPE package

Specifications

Capacity (English) 0.34 oz.
Certifications/Compliance DNase, RNase, and Pyrogen free quality certificate available upon request, Traceability Certificate available upon request
Color White
Capacity (Metric) 10 mL
Material Polystyrene
Sterility Non-sterile
Type Reagent Reservoir
For Use With (Application) Pipetting procedures
Packaging Bulk Wrapped
Quantity 100 Pk.
Autoclavable Not Autoclavable
Channels 1
Disposable Single-use
Bottom Shape V-bottom
Show More Show Less