missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ReadyProbes™ Reagent F-Actin Phalloidin Conjugates

Catalog No. p-200102227
Encompass
Change view
Click to view available options
Color:
Green
Red-Orange
Excitation Wavelength Range:
495/518
540/565
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Excitation Wavelength Range
R37110 Green 495/518
R37112 Red-Orange 540/565
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R37110 Supplier Invitrogen™ Supplier No. R37110
Only null left

Includes

2 x 2.5 mL dropper bottles

Perfect for fixed cells and tissue, ReadyProbes Reagent F-actin phalloidin conjugates offer clear visualization of actin filaments, making it perfect for cell biology, neuroscience, cancer research, and more.

ActinGreen 488 ReadyProbes Reagent is a selective, high-affinity F-actin probe conjugated to our bright, photostable, green-fluorescent Alexa Fluor 488 dye. ActinGreen 488 reagent is a room temperature-stable solution of Alexa Fluor 488 Phalloidin (Cat. No. A12379) packaged in a dropper bottle. ActinRed 555 ReadyProbes Reagent is a comparable F-actin stain conjugated to the red-orange fluorescent dye tetramethylrhodamine (TRITC). ActinRed 555 reagent is a room temperature-stable solution of Rhodamine Phalloidin (Cat. No. R415) packaged in a dropper bottle.

These ready-to-use solutions combines superior brightness with exceptional F-actin staining. Phalloidin is a bi-cyclic peptide, commonly used in imaging applications to selectively label F-actin. Fluorescently labeled phalloidin has several advantages over antibodies for actin labeling, including virtually identical binding properties with actin from different species of plants and animals and low non-specific binding.

Features

  • High-affinity staining of F-actin with superior specificity compared to antibody methods
  • ActinGreen 488 Excitation/Emission: (495/518 nm), detected through standard GFP and FITC filter sets
  • ActinRed 555 Excitation/Emission: (540/565 nm), detected through standard RFP and TRITC filters sets
  • Use counterstain (i.e., DAPI or NucBlue ReadyProbes reagent) as desired

ReadyProbes format advantages

  • Ready-to-use formulation—no need to dilute, weigh, or pipette
  • Just use two drops per mL of medium to stain, incubate 30 minutes, wash, then image
  • Stable at room temperature—keep handy on the benchtop

Applications

  • For fixed and permeabilized cells or tissue
  • Fluorescence microscopy (IF/IHC/ICC)
  • High content screening, high content imaging
  • Flow cytometry

Suggestions for use

  • Both ActinRed 555 and ActinGreen 488 reagents may be added directly to fixed cells in full medium or buffer solutions
  • In most cases 2 drops/mL and an incubation time of 15–30 minutes is sufficient for bright actin staining; however, optimization may be needed for some cell types, conditions, and applications. In such cases, simply add more or fewer drops until the optimal staining intensity is obtained.

Specifications

Color Green
Content And Storage

2 x 2.5 mL dropper bottles

Store at ≤ 25°C.

Excitation Wavelength Range 495/518
Dye Type Alexa Fluor™ 488
For Use With (Equipment) Confocal Microscope, Floid™ Cell Imaging System, Fluorescence Microscope, Flow Cytometer, Floid™ Cell Imaging System
Quantity 1 kit
Detection Method Fluorescence
Description ActinGreen™ 488 ReadyProbes™ Reagent (AlexaFluor™ 488 phalloidin)
Emission Visible
Form Liquid
Product Line ActinGreen, ReadyProbes
Technique Fluorescence Intensity
Label Type Fluorescent Dye
Product Type Phalloidin
Sub Cellular Localization Cytoskeleton
Show More Show Less
Can I use the ReadyProbes reagents for flow cytometry?

This is not recommended. The ReadyProbes reagents were developed for imaging applications whereas the Ready Flow reagents were optimized for flow cytometry.

For Research Use Only. Not for use in diagnostic procedures.