missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ReadyProbes™ Hydrophobic Barrier Pap Pen

Catalog No. R3777
Change view
Click to view available options
Quantity:
2 Pens (6 mL Each)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R3777 2 Pens (6 mL Each)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R3777 Supplier Thermo Scientific™ Supplier No. R3777
  • ONLINE EXCLUSIVE PRICE
    $361.00
    $296.02 / Each

    Save $64.98

    18.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The ReadyProbes Hydrophobic Barrier Pap Pen creates a water-repellent barrier when used on a microscope slide or coverslip.

  • The ReadyProbes Hydrophobic Barrier Pap Pen creates a water-repellent barrier when used on a microscope slide or coverslip
  • By lightly drawing a rectangle or circle around the tissue section or cells on the microscope slide, aqueous incubation reagents can be kept inside the circle or rectangle
  • Save precious reagents by incubating with less volume
  • The pen can also be used to keep reagents separate when more than one tissue section is mounted on a microscope slide
  • The light blue barrier line is heat resistant and compatible with most immunohistochemistry (IHC), immunocytochemistry (ICC), immunofluorescence (IF), and in situ hybridization (ISH) reagents, including detergent-containing reagents
  • It is insoluble in alcohol and acetone, but can be removed with clearing agent (e.g., xylene)
  • When applied as directed, the nib provided with each pen will create a 3 mm wide line
  • The pen design ensures the nib can move and that the nib is not adhered to the pen itself
  • In some instances, during shipping, the nib may become dislodged and fall out. Simply put the nib back into the neck of the pen and use as per the supplied instructions
  • The pen residue does fluoresce – apply the Hydrophobic Barrier a few mm away from tissue section or cells, so it does not interfere with imaging

How to Use

  • Dab the nib of the pen on a paper towel, to make sure the flow of the hydrophobic fluid is uninterrupted
  • Lightly trace a circle or rectangle around the tissue/cells 2-4 times, with a thick line, while keeping a few millimeters distance from the edge of the tissue/cells
  • Let the line dry for 20-30 minutes at room temperature
  • Repeat the process, if needed

Specifications

Quantity 2 Pens (6 mL Each)
For Use With (Application) Immunohistochemistry (IHC), Immunocytochemistry (ICC), Immunofluorescence (IF)
Product Line ReadyProbes
Content And Storage Store at room temperature, away from sunlight, or other heat sources.
Shipping Condition Room Temperature
What is the volume for one drop from the ReadyProbes dropper bottles?

The volume for one drop from the ReadyProbes dropper bottles is approximately 41 µL.