missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ReadyLabel™ Antibody Labeling Kits

Catalog No. p-200004743
Change view
Click to view available options
Quantity:
5 x 100 μg of IgG
5 x 20 μg
5 x 20 μg of IgG
Label or Dye:
Alexa Fluor 420
Alexa Fluor 488
Alexa Fluor 514
Alexa Fluor 594
Alexa Fluor 647
Alexa Fluor 700
Alexa Fluor Plus 555
Alexa Fluor Plus 750
Biotin
None
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Label or Dye
R10709 5 x 20 μg of IgG Alexa Fluor 488
R10712 5 x 100 μg of IgG Alexa Fluor 488
R10710 5 x 20 μg of IgG Alexa Fluor 647
R10713 5 x 100 μg of IgG Alexa Fluor 647
R10711 5 x 20 μg of IgG Biotin
R10714 5 x 100 μg of IgG Biotin
R10701 5 x 20 μg of IgG None
R10702 5 x 100 μg of IgG None
R10720 5 x 20 μg Alexa Fluor 514
R10721 5 x 20 μg Alexa Fluor Plus 555
R10722 5 x 20 μg Alexa Fluor 594
R10723 5 x 20 μg Alexa Fluor 700
R10724 5 x 20 μg Alexa Fluor Plus 750
R10725 5 x 20 μg Alexa Fluor 420
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. R10709 Supplier Invitrogen™ Supplier No. R10709
Only null left

ReadyLabel Antibody Labeling Kits enable fast, convenient labeling of off-the-shelf antibodies and cell culture supernatants without prior purification of the antibody solution.

ReadyLabel Antibody Labeling Kits enable fast, convenient labeling of off-the-shelf antibodies and cell culture supernatants without prior purification of the antibody solution. Each kit contains everything needed to perform 5 labeling reactions and will produce purified antibody conjugates, without the purification step.

Each ReadyLabel Antibody Labeling Kit, 100 μg contains everything needed to perform 5 labeling reactions and will produce purified antibody conjugates, without the purification step. The kit can be used to label antibodies in solutions containing BSA or other stabilizing proteins and antibodies in a primary amine-containing buffer such as Tris or glycine, or even cell culture supernatants, without prior purification of the antibody-containing solution. A reactive dye moiety efficiently binds to primary amines of antibodies to form stable, covalently bound antibody-dye conjugates.

Alexa Fluor dyes produce antibody conjugates that are typically brighter with better photostability and lower pH sensitivity relative to traditional dyes (e.g., FITC). Unlike other dyes, the fluorescence of Alexa Fluor conjugates are insensitive to pH between 4 and 10. Alexa Fluor-labeled antibodies can be used for applications including microscopy, flow cytometry, western blotting, ELISA, and indirect FISH.

ReadyLabel Antibody Labeling Kits offer the following benefits:

  • Convenience—Labeling takes one hour, with less than 20 minutes of hands-on time
  • Versatility—Labelling can be performed on IgG antibodies, regardless of species, isotype, or storage buffer
  • Customization—ReadyLabel Flex Antibody Labeling Kits are designed for use with a wide range of amine-reactive labels including Alexa Fluor dyes

Specifications

For Use With (Application) Antibody Production and Labeling
Product Type Antibody Labeling Kit
Quantity 5 x 20 μg of IgG
For Use With (Equipment) Flow Cytometer, Fluorescence Microscope
Product Line ReadyLabel
Chemical Reactivity Amine
Solubility DMSO (Dimethylsulfoxide)
Label or Dye Alexa Fluor 488
Color Green

For Research Use Only.