Learn More
Thermo Scientific™ Gas Regulators for Reacti-Vap™ Evaporators
Catalog No. p-6827003
Change view
Click to view available options
Includes:
1/4 in. Tube Fitting
1/8 in. Tube Fitting
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
03100090 1/8 in. Tube Fitting
03100091 1/4 in. Tube Fitting
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03100090 Supplier Thermo Scientific™ Supplier No. 60181625
Only null left
Use these gas regulators to deliver the right amount of gas for sample evaporation using the Thermo Scientific™ ReactiVap™ Evaporator.
Deliver the right amount of gas to your Thermo Scientific™ ReactiVap™ Evaporator using these gas regulators. The Reacti-Vap Evaporator delivers pressurized gas to easily and efficiently evaporate samples for gas chromatography (GC) analysis. Choose between gas regulators with a 1/8 inch or a 1/4 inch tube fitting.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
For Use With Reacti-Vap Evaporators
Includes 1/8 in. Tube Fitting
Product Type Gas Regulator
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.