Learn More
Thermo Scientific™ Reacti-Therm™ Thermometers
Use these thermometers to measure temperature in dry block heaters of Thermo Scientific™ Reacti-Therm™ Heating and Stirring module.
$685.58 - $702.88
Specifications
Type Thermometer
For Use With (Equipment) Dry Block Heaters and Other Instruments
View More Specs
Products 2
Catalog Number Mfr. No. Operating Temperature (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Operating Temperature (Metric) Price Quantity & Availability  
PI18914
View Documents Promotion Details
Thermo Scientific™
TS18914
0°C to 100°C
Each for $702.88
Only null left
PI18915
View Documents Promotion Details
Thermo Scientific™
TS18915
0°C to 200°C
Each for $685.58
Only null left
Description
Description
Measure temperature in dry block heaters of the Thermo Scientific™ Reacti-Therm™ Heating and Stirring module and other laboratory equipment using Thermo Scientific™ Reacti-Therm™ Thermometers. The thermometers are mercury-free and alcohol-filled for greater safety. PTFE coating ensures that glass is impervious to corrosive materials. They are available in two different temperature ranges.
Specifications
Specifications
Thermometer
Dry Block Heaters and Other Instruments
Videos
SDS
Documents
Documents
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.