Learn More
Thermo Scientific™ Reacti-Therm™ Thermometers
Catalog No. PI18914
Change view
Click to view available options
Operating Temperature (Metric):
0°C to 100°C
0°C to 200°C
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Operating Temperature (Metric)
PI18914 0°C to 100°C
PI18915 0°C to 200°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI18914 Supplier Thermo Scientific™ Supplier No. TS18914
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Use these thermometers to measure temperature in dry block heaters of Thermo Scientific™ Reacti-Therm™ Heating and Stirring module.
Measure temperature in dry block heaters of the Thermo Scientific™ Reacti-Therm™ Heating and Stirring module and other laboratory equipment using Thermo Scientific™ Reacti-Therm™ Thermometers. The thermometers are mercury-free and alcohol-filled for greater safety. PTFE coating ensures that glass is impervious to corrosive materials. They are available in two different temperature ranges.
Specifications
Type Thermometer
Operating Temperature (Metric) 0°C to 100°C
For Use With (Equipment) Dry Block Heaters and Other Instruments
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.