Learn More
RD Plastics Reclosable Poly Bags
Clear, resealable polyethylene bags
Supplier:  RD Plastics C46
 View more versions of this product
Catalog No. 23700239
Only null left
Description
Description
  • Thickness: 2 or 4mil
  • Seal securely with zipper lock
Specifications
Chemical Identifiers
Clear
Provides protection for contents and clear visibility
Polyethylene
Reclosable Poly Bag
Specifications
Clear
Polyethylene
18 x 15 in. (45.7 x 38.1cm)
15 in.
45.7 cm
0.101 mm
Provides protection for contents and clear visibility
4 mil
18 in.
Reclosable Poly Bag
38.1 cm
Product Suggestions
Product Suggestions