missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RD Plastics Clear Reclosable Zip Bags
For non-biohazard use
$137.53 - $258.91
Specifications
| Color | Clear |
|---|---|
| For Use With (Application) | Non-biohazard use, general purpose |
| Material | Polyethylene |
| Catalog Number | Mfr. No. | Dimensions (L x W) | Thickness (English) | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Dimensions (L x W) | Thickness (English) | Price | Quantity & Availability | ||||
|
01818101
|
RD Plastics
C 26 |
30 x 37.5 cm (12 x 15 in.) | 4 mil |
Case of 500 Each for $253.00
|
|
||||
|
191306043
|
RD Plastics
A29 |
20.32 x 30.32 cm (8 x 10 in.) | 2 mil |
Case of 1000 Each for $137.53
|
|
||||
|
19165620
|
RD Plastics
C29 |
30.4 x 30.4 cm (12 x 12 in.) | 4 mil |
Case of 500 Each for $216.82
|
|
||||
|
01817100
|
RD Plastics
A33 |
30 x 37.5 cm (12 x 15 in.) | 2 mil |
Case of 1000 Each for $258.91
|
|
||||
Description
- Constructed of heavy weight 4 mil polyethylene
- Feature reclosable zip seal
Specifications
| Clear | |
| Non-biohazard use, general purpose | |
| Polyethylene |