missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RD Plastics Clear Reclosable Zip Bags
For non-biohazard use
Supplier: RD Plastics C29
Description
- Constructed of heavy weight 4 mil polyethylene
- Feature reclosable zip seal
Chemical Identifiers
| Clear | |
| Non-biohazard use, general purpose | |
| Polyethylene |
Specifications
| Clear | |
| Polyethylene | |
| 30.4 x 30.4 cm (12 x 12 in.) | |
| 12 in. | |
| 30.4 cm |
| Non-biohazard use, general purpose | |
| 4 mil | |
| 12 in. | |
| Reclosable Bag | |
| 30.4 cm |