missing translation for 'onlineSavingsMsg'
Learn More

RBM26 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501559797
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501559797 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501559797 Supplier Bethyl Laboratories Inc Supplier No. A301215A

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Mouse

6 isoforms of the human protein are produced by alternative splicing. Ota T., Nat. Genet. 36:40-45(2004). Dunham A., Nature 428:522-528(2004). Eichmueller S., Proc. Natl. Acad. Sci. U.S.A. 98:629-634(2001).

Specifications

Antigen RBM26
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/ml
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene RBM26
Gene Accession No. Q5T8P6
Gene Alias ARRS2, C13orf10, RP11-255E21.1, SE70-2, ZC3H17, CTCL tumor antigen se70-2, RNA-binding motif protein 26, RNA-binding protein 26, acidic rich RS domain containing 2, cutaneous T-cell lymphoma tumor antigen se70-2
Gene Symbols RBM26
Host Species Rabbit
Immunogen Between 200 and 250
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64062
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less