missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals RBC lysis buffer for mouse

Catalog No. p-7082302
Change view
Click to view available options
Quantity:
250mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62150AK 250mL
AAJ62150AP 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62150AK Supplier Thermo Scientific Chemicals Supplier No. J62150AK
Only null left

For lysis of mouse red blood cells

RBC Lysis Buffer has been reported for use in flow cytometric analysis and cell culture. Red Blood Cell Lysing Buffer has been developed for use in hybridoma protocols to remove red blood cells from mouse splenocyte suspensions before fusion. It is also useful in systems where it may be desirable to remove red blood cells from cell suspensions, such as whole blood.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Quantity 250mL
Format Liquid
Product Type Cell Lysis Reagent
For Use With (Application) For lysis of mouse red blood cells
Content And Storage 2° to 8°C

RUO – Research Use Only