missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals RBC lysis buffer for human

Catalog No. p-7082341
Change view
Click to view available options
Quantity:
250mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62990AK 250mL
AAJ62990AP 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62990AK Supplier Thermo Scientific Chemicals Supplier No. J62990AK
Only null left

For lysis of human red blood cells

For lysis of human red blood cells and can be used to lyse reticulocytes, erythrocytes, and mature RBCs.

RBC Lysis Buffer for human is used in the isolation of deoxyribonucleic acid (DNA) and ribonucleic acid (RNA).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Quantity 250mL
Format Liquid
Product Type Cell Lysis Reagent
For Use With (Application) For lysis of human red blood cells.
Content And Storage 2° to 8°C

RUO – Research Use Only