missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ RAW Cell Lysate
Description
The RAW Cell Lysate is derived from mouse. It has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Cell Line | RAW cell line lysate (Abelson leukemia virus transformed murine macrophage cell line) |
| Quantity | 0.1 mg |
| Storage Buffer | 1X SDS sample buffer: 60mM Tris-HCl (pH 6.8) 10mM EDTA 2 % SDS 0.03% Bromophenol Blue 10% glycerol 0.3% Beta-mercaptoethanol |
| Host Species | Mouse |
| Content And Storage | -80°C in the dark. Avoid freeze-thaw cycles. |
| Product Line | Novus |
For Research Use Only