Learn More
Invitrogen™ Rat Set
Stealth RNAi Predesigned siRNAs are designed to provide robust and long-lasting gene silencing with minimal off-target effects. Stealth RNAi siRNAs are modified synthetic 25-mer RNA duplexes. Our proprietary technology ensures high specificity and stability, making Stealth RNAi siRNAs an ideal choice for researchers seeking reliable and efficient gene knockdown in human, mouse, or rat.
$488.00 - $1,068.00
Specifications
Content And Storage Store at room temperature.
View More Specs
Products 2
Catalog Number Mfr. No. Target Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Target Quantity Price Quantity & Availability  
1320001
Encompass
View Documents Promotion Details
Invitrogen™
LS1320001
Mouse Single 20 nmol
Each for $488.00
Only null left
1320003
Encompass
View Documents Promotion Details
Invitrogen™
LS1320003
Mouse Set 20 x 3 nmol
Each for $1,068.00
Only null left
Description
Description
Stealth RNAi Predesigned siRNAs enable robust and long-lasting gene silencing with reduced off-target effects. Stealth RNAi siRNAs are modified synthetic 25-mer RNA duplexes. Our proprietary technology helps ensure high specificity and stability, making Stealth RNAi siRNAs an excellent choice for researchers seeking reliable and efficient gene knockdown in human, mouse, or rat.
Key features include:
Higher specificity and potency—algorithm design helps reduce off-target effects and increase target gene knockdown
Enhanced stability—engineered for enhanced stability in serum and cellular environments, helping ensure prolonged gene silencing
Limits cellular toxicity—designed to evade immune detection, for reduced risk of immune response
Specifications
Specifications
Store at room temperature.