Learn More
Invitrogen™ Rat SCF ELISA Development Kit (ABTS), PeproTech®
ELISA
Supplier: Invitrogen™ 900K258K
Description
Stem cell factor (SCF) is the ligand of the c-Kit oncogene and is expressed by various structural and inflammatory cells in the airways. Binding of SCF by the c-Kit receptor leads to homodimerization of the receptor and the activation of signalling pathways such as PI-3, PLC-gamma, Jak/STAT, and MAP kinase pathways. SCF expression leads to the induction of mast cell survival and the expression and release of histamine, pro-inflammatory cytokines and chemokines. The inhibition of the SCF/c-Kit pathway leads to a decrease in histamine levels, mast cell and eosinophil infiltration, IL-4 production and airway hyperresponsiveness, suggesting this pathway may be a useful therapeutic target in inflammatory diseases such as asthma. At least two isoforms of SCF are known to exist.Chemical Identifiers
| Lot-specific | |
| Lot-specific | |
| Colorimetric Microplate Reader |
Specifications
| Lot-specific | |
| HRP | |
| Colorimetric Microplate Reader | |
| HRP | |
| RUO | |
| SCF | |
| 24 hrs 30 mins |
| Lot-specific | |
| ELISA | |
| Capture Antibody, Detection Antibody, Protein Standard, HRP-conjugate | |
| 10 x 96 Tests | |
| Refer to product documentation for component specific storage temperature. | |
| 1 hrs 45 mins |