missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat Jagged 1 ELISA Kit

ELISA

Specifications

Accession Number ABE2; AGS; AHD; AWS; CD339; Gsfabe2; HJ1; Htu; JAG1; jagged 1; jagged canonical Notch ligand 1; Jagged1; JAGL1; Ozz; protein jagged-1; Ser-1; Serrate-1
Assay Range 73 to 18,000 pg/mL
Assay Sensitivity 73 pg/mL
Conjugate HRP
Product Type ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ERA35RBX5
Encompass
View Documents Promotion Details
Invitrogen™
ERA35RBX5
5 x 96 Tests N/A
Only null left
 
ERA35RB
Encompass
View Documents Promotion Details
Invitrogen™
ERA35RB
96 Tests N/A
Only null left
 
Description

Description

Rat Jagged 1 quantitates rat Jagged 1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rat Jagged 1.

The jagged 1 protein encoded by JAG1 is the human homolog of the Drosophilia jagged protein. Human jagged 1 is the ligand for the receptor notch 1, the latter a human homolog of the Drosophilia jagged receptor notch. Mutations that alter the jagged 1 protein cause Alagille syndrome. Jagged 1 signalling through notch 1 has also been shown to play a role in hematopoiesis.
Specifications

Specifications

ABE2; AGS; AHD; AWS; CD339; Gsfabe2; HJ1; Htu; JAG1; jagged 1; jagged canonical Notch ligand 1; Jagged1; JAGL1; Ozz; protein jagged-1; Ser-1; Serrate-1
73 pg/mL
ELISA
Colorimetric Microplate Reader
JAG1
<10
HRP
Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Jagged1
4 hrs 45 mins
73 to 18,000 pg/mL
HRP
Plasma, Serum, Supernatant
29146
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
RUO
2°C to 8°C
1 hrs 20 mins
Videos
SDS
Documents

Documents