Learn More
Invitrogen™ Rat Insulin ELISA Kit
Description
The Rat Insulin ELISA quantitates Rt Insulin in rat serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Rt Insulin. Principle of the method The Rat Insulin solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Specifications
| Accession Number | P01322, P01323 |
| Assay Range | 5-300 μIU/mL |
| Assay Sensitivity | 5 μIU/mL |
| Calibration Range | 5 μIU/mL to 300 μIU/mL |
| Content And Storage | Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability. |
| Description | Insulin ELISA Kit, 96 Assays, 1 Kit, Rat Species, Hormones and Receptors Protein Family, Colorimetric Detection Method, Enzyme-Linked Immunosorbent Assay Designed for the Quantitation and Detection of Insulin in Samples |
| Detectable Analytes | Rat, mouse, human, porcine and bovine insulin and proinsulin, rat insulin in serum, plasma, and cell culture media |
| Assay | Solid-phase sandwich ELISA |
| Conjugate | HRP |
| Detection Method | Colorimetric |
| Show More |