missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat IL-10 ELISA Kit

Catalog No. p-7219890
Encompass
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ERA23RBX5 5 x 96 Tests
ERA23RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ERA23RBX5 Supplier Invitrogen™ Supplier No. ERA23RBX5
Only null left

ELISA

Rat IL-10 quantitates rat IL-10 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rat IL-10.

IL-10 encodes a protein that acts as a cytokine and is primarily produced by monocytes, with some production by lymphocytes. This cytokine has various effects on immunoregulation and inflammation, including down-regulating the expression of Th1 cytokines, MHC class II Ags, and costimulatory molecules on macrophages. It also enhances B cell survival, proliferation, and antibody production. Additionally, IL-10 can block NF-kappa B activity and is involved in the regulation of the JAK-STAT signaling pathway. Knockout studies in mice have suggested that this cytokine is an essential immunoregulator in the intestinal tract. Mutations in this gene have been associated with an increased susceptibility to HIV-1 infection and rheumatoid arthritis. Diseases associated with IL10 include Graft-Versus-Host Disease and Human Immunodeficiency Virus Type 1.

Specifications

Accession Number CSIF; cytokine; cytokine synthesis inhibitory factor; GVHDS; H-IL-10; il 10; IL10; IL-10; IL-10 precursor; IL10A; IL10X; ILN; Interleukin; interleukin 10; Interleukin10; Interleukin-10; MGC126450; MGC126451; RP11-262N9.1; T-cell growth inhibitory factor; TGIF
Assay Range 10 to 6,000 pg/mL
Assay Sensitivity 10 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 25325
Gene Symbol IL10
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target IL-10
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less