missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Accession Number | AW554192; GAS1; Gas-1; growth arrest specific 1; growth arrest-specific 1; Growth arrest-specific gene-1; growth arrest-specific protein 1 |
|---|---|
| Assay Range | 0.05 to 10 ng/mL |
| Assay Sensitivity | 0.05 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ERA18RBX5
|
Invitrogen™
ERA18RBX5 |
5 x 96 Tests |
Each for $4,420.00
|
|
|||||
ERA18RB
|
Invitrogen™
ERA18RB |
96 Tests |
Each for $938.00
|
|
|||||
Description
Rat Gas 1 quantitates rat Gas 1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rat Gas 1.
Growth arrest-specific 1 plays a role in growth suppression. GAS1 blocks entry to S phase and prevents cycling of normal and transformed cells. Gas1 is a putative tumor suppressor gene.Specifications
| AW554192; GAS1; Gas-1; growth arrest specific 1; growth arrest-specific 1; Growth arrest-specific gene-1; growth arrest-specific protein 1 | |
| 0.05 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| GAS1 | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| GAS1 | |
| 4 hrs 45 mins |
| 0.05 to 10 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 683470 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |