missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat Calcitonin ELISA Kit

Catalog No. EEL226
Change view
Click to view available options
:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
EEL226 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEL226 Supplier Invitrogen™ Supplier No. EEL226
Only null left

ELISA

Calcitonin is a 32 amino acid peptide which can be demonstrated in C cells of the normal and hyperplastic thyroid. Staining for calcitonin may be used for the identification of a spectrum of C cell proliferative abnormalities ranging from C cell hyperplasia to invasive tumors. Staining for calcitonin in medullary carcinoma of the thyroid produces a fine granular pattern in the cytoplasm. Amyloid deposits within the tumor may also exhibit varying degrees of calcitonin activity.

Specifications

Accession Number P01256, P01257
Assay Range 15.63 to 1000 pg/mL
Assay Sensitivity 9.38 pg/mL
Conjugate HRP
Detection Method Colorimetric
Immunoassay Kit Format ELISA Microplate (Dismountable)
Product Type ELISA
Sample Type Plasma, Serum
Target Species Rat
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 24241
Gene Symbol CALCA
Interassay CV <10
Intraassay CV <10
Kit Contents Micro ELISA Plate (Dismountable), Reference Standard, Concentrated Biotinylated Detection Ab(100x), Concentrated HRP Conjugate (100x), Reference Standard & Sample Diluent, Biotinylated Detection Ab Diluent, HRP Conjugate Diluent, Concentrated Wash Buffer(25x), Substrate Reagent, Stop Solution, Plate Sealer, Product Description, Certificate of Analysis
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma: 100 μL, Serum: 100 μL
Storage Requirements Refer to product documentation for component specific storage temperature.
Target Calcitonin
Test Time 1 hr. 20 min.
Total Assay Time 3 hr. 30 min.
Show More Show Less