missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat anti-Mouse Lambda Light Chain Secondary Antibody, PE

Catalog No. 501137526
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501137526 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501137526 Supplier Invitrogen™ Supplier No. MLAMBDA04

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rat Monoclonal Secondary Antibody

Rat anti-Mouse Lambda Light Chain Secondary Antibody for Flow, ELISA

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Mouse Lambda Light Chain, Rat anti-Mouse Lambda Light Chain Secondary Antibody, PE
Applications ELISA, Flow Cytometry
Classification Monoclonal
Clone JC5-1
Concentration 0.1 mg/mL
Conjugate PE
Formulation PBS with <0.1% sodium azide; pH 7.5
Gene Alias Immunoglobulin lambda light chain, Ig lambda light chain, Ig lambda, lambda light chain, Igl
Host Species Rat
Immunogen Mouse Lambda light chain.
Purification Method purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Secondary Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less