missing translation for 'onlineSavingsMsg'
Learn More

Rat Anti-Mouse IgG2B Fluorescein-conjugated Antibody, R&D Systems™

Catalog No. F0134
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0134 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F0134 Supplier Bio-Techne Supplier No. F0134

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rat Monoclonal Antibody

IgG2b Monoclonal specifically detects IgG2b in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen IgG2b
Applications Flow Cytometry
Classification Monoclonal
Clone 332723
Conjugate Fluorescein
Dilution Flow Cytometry
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Host Species Rat
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8degrees C. Do not use past expiration date. Protect from light.
Product Type Antibody
Isotype IgG1
Product Line R&D Systems
Show More Show Less