missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat anti-Mouse IgG2a Secondary Antibody, HRP

Catalog No. 01672169
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01672169 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01672169 Supplier Invitrogen™ Supplier No. SA135646
  • ONLINE EXCLUSIVE PRICE
    $3,850.85
    $2,238.75 / Each

    Save $1,612.10

    42.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rat Monoclonal Secondary Antibody

Mouse gamma 2a heavy chain of immunoglobulin (determined by immunodot). No crossreactivity with other animals. Useful in immunoassays, such as ELISA's (good binding on plastic), immunohistochemistry on frozen sections and for affinity chromatog¬raphy of murine IgG1. Available unconjugated MON5068, HRP conjugated 5068H.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Mouse IgG2a
Applications ELISA, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone LO-MG2a-9
Concentration 1 mg/mL
Conjugate HRP
Formulation PBS with 50% glycerol and no preservative
Host Species Rat
Immunogen Immunoglobulins.
Purification Method Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage -20°C
Product Type Secondary Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less