missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat anti-Mouse IgG2a Secondary Antibody, eFluor™ 570, eBioscience™

Catalog No. 41421080
Encompass
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41421080 25 μg
41421082 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 41421080 Supplier Invitrogen™ Supplier No. 41421080

Rat Monoclonal Secondary Antibody

Description: The monoclonal antibody m2a-15F8 recognizes Mouse IgG2a antibodies and can be used as a second step reagent in microscopy. The monoclonal does not recognize other mouse isotypes nor does it crossreact to rat IgG2a or any rat isotype antibodies. Applications Reported: This m2a-15F8 antibody has been reported for use in immunohistochemical staining of frozen tissue sections, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, immunohistochemical staining, and immunocytochemistry. Applications Tested: This m2a-15F8 antibody has been tested immunocytochemistry and immunohistochemistry after staining with a primary antibody of the mouse IgG2a isotype. This can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. This product has not been validated for flow cytometric analysis. Filter Recommendation: When using this eFluor® 570 antibody conjugate, we recommend a filter that will capture the 570 emission wavelength (for example, Excitation 545/25, 565LP, Emission 605/70). A standard Alexa Fluor® 555 or TRITC filter is acceptable. Excitation: 555 nm; Emission: 570 nm.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Mouse IgG2a
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Monoclonal
Clone m2a-15F8
Concentration 0.2 mg/mL
Conjugate eFluor 570
Formulation PBS with 0.09% sodium azide; pH 7.2
Host Species Rat
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Secondary Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less