missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat anti-Mouse IgG2a Secondary Antibody

Catalog No. 046200
Encompass
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
046200 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 046200 Supplier Invitrogen™ Supplier No. 046200

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rat Monoclonal Secondary Antibody

This monoclonal Rat anti-Mouse IgG2a reacts with mouse IgG2a and exhibits no reactivity to the other mouse IgG subclasses, IgA, IgM or light chains.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Mouse IgG2a
Applications ELISA, Western Blot
Classification Monoclonal
Clone LO-MG2a-3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Host Species Rat
Immunogen Mouse IgG2a.
Purification Method Purified
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage -20°C
Product Type Secondary Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less