missing translation for 'onlineSavingsMsg'
Learn More

Rat anti-Mouse IgG2a Heavy Chain Secondary Antibody (LO-MG2a-7) [Janelia Fluor 525], R&D Systems™

Catalog No. NB786027
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB786027 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB786027 Supplier Bio-Techne Supplier No. NBP268435JF525

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems. Rat Monoclonal Antibody

IgG2a Heavy Chain Monoclonal antibody specifically detects IgG2a Heavy Chain in Mouse samples. It is validated for ELISA

Specifications

Antigen IgG2a Heavy Chain
Applications ELISA
Classification Monoclonal
Clone LO-MG2a-7
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Host Species Rat
Immunogen Purified IgG from BALB/c mice
Purification Method Affinity purified
Quantity 0.25 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less