Learn More
Invitrogen™ Rat anti-Mouse IgG1 Secondary Antibody, Super Bright™ 780, eBioscience™
Description
Description: The monoclonal antibody M1-14D12 recognizes mouse IgG1 antibodies and can be used as a second step reagent in flow cytometry and microscopy. The monoclonal does not recognize other mouse isotypes nor does it crossreact to rat IgG1 or any rat isotype antibodies. Applications Reported: This M1-14D12 antibody has been reported for use in flow cytometric analysis. Applications Tested: This M1-14D12 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 780 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 780 nm. We recommend using a 780/60 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product # SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. In some experiments, we have observed that compensation values for Super Bright 780-conjugated antibodies are higher in the violet 450/50 channel when using UltraComp eBeads microspheres (Product #...
Spécifications
Spécifications
| Antigène | Mouse IgG1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | M1-14D12 |
| Concentration | 0.2 mg/mL |
| Conjugué | Super Bright 780 |
| Formule | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Espèces hôtes | Rat |
| Méthode de purification | Affinity chromatography |
| Quantité | 100 μg |
| Afficher plus de résultats |