missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rat anti-Mouse IgG1 Secondary Antibody, HRP
Description
Mouse gamma 1 heavy chain of immunoglobulin (determined by immunodot). No crossreactivity with other animals. Available as unconjugated MON5053, FITC conjugated MON 5053F, HRP conjugated Cat.no. MON 5053P, Biotin conjugated MON 5053B.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Spécifications
Spécifications
| Antigène | Mouse IgG1 |
| Applications | ELISA, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | LO-MG1-2 |
| Concentration | 1 mg/mL |
| Conjugué | HRP |
| Formule | PBS with 0.1% sodium azide |
| Espèces hôtes | Rat |
| Immunogène | Proprietary Immunogen. |
| Méthode de purification | Purified |
| Afficher plus de résultats |