Learn More
Invitrogen™ Rat anti-Mouse IgG1 Secondary Antibody, Biotin, eBioscience™
Description
Description: The monoclonal antibody M1-14D12 recognizes Mouse IgG1 antibodies and can be used as a second step reagent in flow cytometry and microscopy applications. The monoclonal does not recognize other mouse isotypes nor does it crossreact to rat IgG1 or any rat isotype antibodies. Applications Reported: This M1-14D12 antibody has been reported for use in flow cytometric analysis, immunohistochemical staining of frozen tissue sections, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, and immunocytochemistry. Applications Tested: This M1-14D12 antibody has been tested by immunocytochemistry on fixed and permeabilized cells using a mouse IgG1 primary antibody. This antibody can be used at less than or equal to 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.
Spécifications
Spécifications
| Antigène | Mouse IgG1 |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Monoclonal |
| Clone | M1-14D12 |
| Concentration | 0.5 mg/mL |
| Conjugué | Biotin |
| Formule | PBS with 0.09% sodium azide; pH 7.2 |
| Espèces hôtes | Rat |
| Méthode de purification | Affinity chromatography |
| Quantité | 100 μg |
| Afficher plus de résultats |