missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat anti-Mouse Kappa Light Chain Secondary Antibody, HRP

Catalog No. MA134732
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA134732 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA134732 Supplier Invitrogen™ Supplier No. MA134732

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rat Monoclonal Secondary Antibody

Does not cross-react with other mouse immunoglobulin classes or subclasses. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Mouse Kappa Light Chain
Applications ELISA, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone LO-MK-1
Concentration 1 mg/mL
Conjugate HRP
Formulation PBS with 50% glycerol and 0.01% thimerosal
Gene Alias Iglc
Host Species Rat
Immunogen Purified mouse IgG1 kappa.
Purification Method Affinity chromatography
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Secondary Antibody
Form Liquid
Isotype IgG2a
Show More Show Less